Order Entry
Italy
Orders LinkContactUsLinkComponent
Vitamin E 99%

Vitamin E 99%

Bottle
  APOSOR1015423-25G
 
New Product
Vitamin E 99%
  APOSOR1015423-25G
New Product
 :  59-02-9
Vitamin E 99%

Bottle

 :  59-02-9
 
30,40 €
   
1 * 25 g
 

Usage: Organic building block/ reagent.

 :  (+)-α-Tocopherol, Vitamin E Alcohol, (R)-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, α-Vitamin E, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E, D(+)-α-Tocopherol
  •  :59-02-9
  •  :C₂₉H₅₀O₂
  •  :430,71 g/mol
  •  :200-412-2
 

 

  • Pack type:
    Bottle

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.