Order Entry
Italy
Orders LinkContactUsLinkComponent
Vitamin E 99%

Vitamin E 99%

Bottle
Catalog # APOSOR1015423-5G
Irritant
New Product
Supplier:   Apollo Scientific
Vitamin E 99%
Catalog # APOSOR1015423-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  59-02-9
Vitamin E 99%

Bottle

CAS Number:  59-02-9
Price
24,60 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  (+)-α-Tocopherol, Vitamin E Alcohol, (R)-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, α-Vitamin E, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E, D(+)-α-Tocopherol
  • CAS Number:59-02-9
  • Formula:C₂₉H₅₀O₂
  • Molecular Weight:430,71 g/mol
  • EINECS:200-412-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.