Order Entry
Italy
Orders LinkContactUsLinkComponent
 

3,4-Dimethylanisole

Bottle
Catalog # APOSOR2170-2.5KG
Corrosive
Irritant
New Product
Supplier:   Apollo Scientific
3,4-Dimethylanisole
Catalog # APOSOR2170-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  4685-47-6
3,4-Dimethylanisole

Bottle

CAS Number:  4685-47-6
Price
2 070,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:4685-47-6
  • Formula:C₉H₁₂O
  • MDL Number:MFCD00008396
  • Molecular Weight:136.19 g/mol
  • Boiling Pt:200…201 °C
  • Density:0.974 g/cm³ (25 °C)
  • Flash Pt:76 °C
  • Storage Temperature:Ambient
  • EINECS:225-142-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications