3,4-Dimethylanisole
Bottle
Catalog # APOSOR2170-100G
New Product
Supplier: Apollo Scientific
3,4-Dimethylanisole
Catalog # APOSOR2170-100G
Supplier: Apollo Scientific
New Product
CAS Number: 4685-47-6
Jump to:
Product Details & Documents
- CAS Number:4685-47-6
- Formula:C₉H₁₂O
- MDL Number:MFCD00008396
- Molecular Weight:136.19 g/mol
- Boiling Pt:200…201 °C
- Density:0.974 g/cm³ (25 °C)
- Flash Pt:76 °C
- Storage Temperature:Ambient
- EINECS:225-142-2
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSOR2170-100G
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



