Order Entry
Italy
Orders LinkContactUsLinkComponent
3,4-Dimethylanisole

3,4-Dimethylanisole

Bottle
Catalog # APOSOR2170-5G
Corrosive
Irritant
New Product
Supplier:   Apollo Scientific
3,4-Dimethylanisole
Catalog # APOSOR2170-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  4685-47-6
3,4-Dimethylanisole

Bottle

CAS Number:  4685-47-6
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:4685-47-6
  • Formula:C₉H₁₂O
  • MDL Number:MFCD00008396
  • Molecular Weight:136.19 g/mol
  • Boiling Pt:200…201 °C
  • Density:0.974 g/cm³ (25 °C)
  • Flash Pt:76 °C
  • Storage Temperature:Ambient
  • EINECS:225-142-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.