Order Entry
Italy
Orders LinkContactUsLinkComponent
 

2-Hydroxy-1-naphthaldehyde

Bottle
Catalog # APOSOR8817-2.5KG
Irritant
Environment
New Product
Supplier:   Apollo Scientific
2-Hydroxy-1-naphthaldehyde
Catalog # APOSOR8817-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  708-06-5
2-Hydroxy-1-naphthaldehyde

Bottle

CAS Number:  708-06-5
Price
808,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  2-Hydroxy-α-naphthaldehyde
  • CAS Number:708-06-5
  • Formula:C₁₁H₈O₂
  • MDL Number:MFCD00004005
  • Molecular Weight:172.18 g/mol
  • Boiling Pt:191…193 °C/27mmHg
  • Melting Pt:76…80 °C
  • Storage Temperature:Ambient
  • EINECS:211-902-0
  • UN:3077
  • ADR:9+EHS,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications