Order Entry
Italy
Orders LinkContactUsLinkComponent
 

3,3-Dimethylcyclohexanone

Bottle
Catalog # APOSOR315523-1G
Flammable
Corrosive
New Product
Supplier:   Apollo Scientific
3,3-Dimethylcyclohexanone
Catalog # APOSOR315523-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  2979-19-3
3,3-Dimethylcyclohexanone

Bottle

CAS Number:  2979-19-3
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:2979-19-3
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00040178
  • Molecular Weight:126.2 g/mol
  • Boiling Pt:175 °C
  • Density:0.91 g/cm³ (25 °C)
  • Flash Pt:59 °C
  • Storage Temperature:Ambient
  • UN:1224
  • ADR:3,III
  •  :Density: 0.909 g/cm³ (25 °C)
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.