Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Hydroxy-4-methyl-3-nitropyridine 98%

2-Hydroxy-4-methyl-3-nitropyridine 98%

Bottle
Codice catalogo APOSOR3150-500G
New Product
Nome del fornitore:   Apollo Scientific
2-Hydroxy-4-methyl-3-nitropyridine 98%
Codice catalogo APOSOR3150-500G
Nome del fornitore:  Apollo Scientific
New Product
Numero CAS:  21901-18-8
2-Hydroxy-4-methyl-3-nitropyridine 98%

Bottle

Numero CAS:  21901-18-8
Price
859,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  4-Methyl-3-nitro-2-pyridone, 2-Hydroxy-4-methyl-3-nitropyridine
  • CAS Number:21901-18-8
  • Formula:C₆H₆N₂O₃
  • MDL Number:MFCD00010689
  • Molecular Weight:154,12344 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications