Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Thioacetamide 98%

Bottle
Catalog # APOSOR28604-2.5KG
Irritant
Health
New Product
Supplier:   Apollo Scientific
Thioacetamide 98%
Catalog # APOSOR28604-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  62-55-5
Thioacetamide 98%

Bottle

CAS Number:  62-55-5
Price
254,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Ethanethioamide
  • CAS Number:62-55-5
  • Formula:CH₃CSNH₂
  • MDL Number:MFCD00008070
  • Molecular Weight:75.13 g/mol
  • Melting Pt:112…114 °C
  • Storage Temperature:Ambient
  • EINECS:200-541-4
Specifications

Specifications

  • Pack type:
    Bottle

Specifications