Order Entry
Italy
Orders LinkContactUsLinkComponent
Strata C18-E SPE Tubes
New Product
Supplier:   PHENOMENEX
Strata C18-E SPE Tubes
Supplier:  PHENOMENEX
New Product
Certificates
Strata C18-E SPE Tubes
Strata C18-E SPE Tubes
33 Options Available Below
Product Details & Documents
Product Family Options
Tubes
Product Information
  • Phase
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C18-E
    PP
    70 Å
    55 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 281,00 €
    281,00 €
    1 * 100 pezzi
       281,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 500 pezzi
    1 * 500 pezzi - 1 076,00 €
    1 076,00 €
    1 * 500 pezzi
       1 076,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 281,00 €
    281,00 €
    1 * 100 pezzi
       281,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    3 ml
    80 mg
    Log in for availability
    Log in for availability
    1 * 50 pezzi
    1 * 50 pezzi - 260,00 €
    260,00 €
    1 * 50 pezzi
       260,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 pezzi
    1 * 50 pezzi - 194,00 €
    194,00 €
    1 * 50 pezzi
       194,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 200 pezzi
    1 * 200 pezzi - 515,00 €
    515,00 €
    1 * 200 pezzi
       515,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 pezzi
    1 * 50 pezzi - 209,00 €
    209,00 €
    1 * 50 pezzi
       209,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 pezzi
    1 * 200 pezzi - 597,00 €
    597,00 €
    1 * 200 pezzi
       597,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 pezzi
    1 * 50 pezzi - 224,00 €
    224,00 €
    1 * 50 pezzi
       224,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 200 pezzi
    1 * 200 pezzi - 668,00 €
    668,00 €
    1 * 200 pezzi
       668,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    6 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 30 pezzi
    1 * 30 pezzi - 173,00 €
    173,00 €
    1 * 30 pezzi
       173,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 pezzi
    1 * 30 pezzi - 173,00 €
    173,00 €
    1 * 30 pezzi
       173,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 200 pezzi
    1 * 200 pezzi - 755,00 €
    755,00 €
    1 * 200 pezzi
       755,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 pezzi
    1 * 30 pezzi - 224,00 €
    224,00 €
    1 * 30 pezzi
       224,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 200 pezzi
    1 * 200 pezzi - 1 076,00 €
    1 076,00 €
    1 * 200 pezzi
       1 076,00 €
    Specifications:
    New Product
    New Product
Tubes with ASPEC cap
Product Information
  • Phase
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C18-E
    PP
    70 Å
    55 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 275,00 €
    275,00 €
    1 * 100 pezzi
       275,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 500 pezzi
    1 * 500 pezzi - 1 107,00 €
    1 107,00 €
    1 * 500 pezzi
       1 107,00 €
    Specifications:
    New Product
    New Product
Teflon tubes
Product Information
  • Phase
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C18-E
    PP
    70 Å
    55 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 pezzi
    1 * 50 pezzi - 250,00 €
    250,00 €
    1 * 50 pezzi
       250,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 pezzi
    1 * 30 pezzi - 214,00 €
    214,00 €
    1 * 30 pezzi
       214,00 €
    Specifications:
    New Product
    New Product
Tabless tubes
Product Information
  • Phase
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C18-E
    PP
    70 Å
    55 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 418,00 €
    418,00 €
    1 * 100 pezzi
       418,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 pezzi
    1 * 50 pezzi - 214,00 €
    214,00 €
    1 * 50 pezzi
       214,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 200 pezzi
    1 * 200 pezzi
    Specifications:
    New Product
    New Product
Tabless tubes with ASPEC cap
Product Information
  • Phase
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C18-E
    PP
    70 Å
    55 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 500 pezzi
    1 * 500 pezzi - 2 213,00 €
    2 213,00 €
    1 * 500 pezzi
       2 213,00 €
    Specifications:
    New Product
    New Product
Centrifuge tubes
Product Information
  • Phase
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C18-E
    PP
    70 Å
    55 µm
    50 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 pezzi
    1 * 50 pezzi - 355,00 €
    355,00 €
    1 * 50 pezzi
       355,00 €
    Specifications:
    New Product
    New Product
Giga tubes
Product Information
  • Phase
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C18-E
    PP
    70 Å
    55 µm
    12 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 20 pezzi
    1 * 20 pezzi - 224,00 €
    224,00 €
    1 * 20 pezzi
       224,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    12 ml
    2000 mg
    Log in for availability
    Log in for availability
    1 * 20 pezzi
    1 * 20 pezzi - 224,00 €
    224,00 €
    1 * 20 pezzi
       224,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    20 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 20 pezzi
    1 * 20 pezzi - 296,00 €
    296,00 €
    1 * 20 pezzi
       296,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    20 ml
    5000 mg
    Log in for availability
    Log in for availability
    1 * 20 pezzi
    1 * 20 pezzi - 408,00 €
    408,00 €
    1 * 20 pezzi
       408,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    60 ml
    10000 mg
    Log in for availability
    Log in for availability
    1 * 16 pezzi
    1 * 16 pezzi - 500,00 €
    500,00 €
    1 * 16 pezzi
       500,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    60 ml
    20000 mg
    Log in for availability
    Log in for availability
    1 * 16 pezzi
    1 * 16 pezzi - 734,00 €
    734,00 €
    1 * 16 pezzi
       734,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    150 ml
    20000 mg
    Log in for availability
    Log in for availability
    1 * 8 pezzi
    1 * 8 pezzi - 1 097,00 €
    1 097,00 €
    1 * 8 pezzi
       1 097,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    150 ml
    50000 mg
    Log in for availability
    Log in for availability
    1 * 8 pezzi
    1 * 8 pezzi - 1 097,00 €
    1 097,00 €
    1 * 8 pezzi
       1 097,00 €
    Specifications:
    New Product
    New Product
  • C18-E
    PP
    70 Å
    55 µm
    150 ml
    70000 mg
    Log in for availability
    Log in for availability
    1 * 8 pezzi
    1 * 8 pezzi - 1 545,00 €
    1 545,00 €
    1 * 8 pezzi
       1 545,00 €
    Specifications:
    New Product
    New Product