Tridecanoic acid-methyl ester
Tridecanoic acid-methyl ester
Catalog # EHERC17818150
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCCCCCCCCCCC(=O)OC
- Inchi: InChI=1S/C14H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-14(15)16-2/h3-13H2,1-2H3
- IUPAC: methyl tridecanoate
Specifications
Specifications
- Description:Tridecanoic acid-methyl ester
- Cat. No.:EHERC17818150
- Formula:C₁₄H₂₈O₂
- Storage conditions:20 °C
- Hazard class description:GHS05, GHS09
- GHS signal:Warning
- Molecular weight:228,37
- CAS No.:1731-88-0
- Shipping conditions:RT



