Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Flucycloxuron

Flucycloxuron
Catalog # EHERC13698500
Supplier:   EHRENSTORFER
Flucycloxuron
Catalog # EHERC13698500
Supplier:  EHRENSTORFER
CAS Number:  
Flucycloxuron
Flucycloxuron

Flucycloxuron

CAS Number:  
Price
319,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
  • SMILES: Fc1cccc(F)c1C(=O)NC(=O)Nc2ccc(CON=C(C3CC3)c4ccc(Cl)cc4)cc2
  • Inchi: InChI=1S/C25H20ClF2N3O3/c26-18-10-8-17(9-11-18)23(16-6-7-16)31-34-14-15-4-12-19(13-5-15)29-25(33)30-24(32)22-20(27)2-1-3-21(22)28/h1-5,8-13,16H,6-7,14H2,(H2,29,30,32,33)
  • IUPAC: N-[[4-[[[(4-chlorophenyl)-cyclopropylmethylidene]amino]oxymethyl]phenyl]carbamoyl]-2,6-difluorobenzamide
  • Packing group: III
Specifications

Specifications

  • Description:
    Flucycloxuron
  • UN No.:
    3077
  • Cat. No.:
    EHERC13698500
  • Formula:
    C₂₅H₂₀ClF₂N₃O₃
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS07, GHS09
  • GHS signal:
    Warning
  • Synonym(s):
    A 1335;Andalin;Andalin DC 25;DU 319722;Flucycloxuron;N-[[[4-[[[[(4-Chlorophenyl)cyclopropylmethylene]amino]oxy]methyl]phenyl]amino]carbonyl]-2,6-difluorobenzamide;OMS 3041;PH 70-23;UBI-A 1335;N-[[[4-[[[[(4-chlorophenyl)cyclopropylmethylene]amino]oxy]methyl]phenyl]amino]carbonyl]-2,6-difluoro-Benzamide
  • Molecular weight:
    483,89
  • CAS No.:
    113036-88-7
  • Shipping conditions:
    RT

Specifications