Order Entry
Italy
Orders LinkContactUsLinkComponent
Dimethirimol

Dimethirimol

Dimethirimol
Catalog # EHERC12690000
Supplier:   EHRENSTORFER
Dimethirimol
Catalog # EHERC12690000
Supplier:  EHRENSTORFER
CAS Number:  
Dimethirimol
Dimethirimol

Dimethirimol

CAS Number:  
Price
345,00 €
Log in for availability
1 * 0,1 g
Product Details & Documents
  • SMILES: CCCCc1c(C)nc(nc1O)N(C)C Inchi: InChI=1S/C11H19N3O/c1-5-6-7-9-8(2)12-11(14(3)4)13-10(9)15/h5-7H2,1-4H3,(H,12,13,15) IUPAC: 5-butyl-2-(dimethylamino)-6-methylpyrimidin-4-ol
  • Packing group: III
Specifications

Specifications

  • Description:
    Dimethirimol
  • UN No.:
    2588
  • Cat. No.:
    EHERC12690000
  • Formula:
    C₁₁H₁₉N₃O
  • Storage conditions:
    20 ℃
  • Hazard class description:
    GHS06, GHS09
  • GHS signal:
    Danger
  • Molecular weight:
    209,29
  • CAS No.:
    5221-53-4
  • Shipping conditions:
    RT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.