Triethylamine
Triethylamine
Catalog # EHERCA17832000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCN(CC)CC
- Inchi: InChI=1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3
- IUPAC: N,N-diethylethanamine
- Packing group: II
Specifications
Specifications
- Description:Triethylamine
- UN No.:1296
- Cat. No.:EHERCA17832000
- Formula:C₆H₁₅N
- Storage conditions:20 °C
- Hazard class description:GHS02, GHS05, GHS06, GHS07, GHS09
- GHS signal:Danger
- Synonym(s):Triethylamine;N,N-Diethylethanamine;(Diethylamino)ethane;N,N,N-Triethylamine;TEA
- Molecular weight:101,19
- CAS No.:121-44-8
- Shipping conditions:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



