Tributyl phosphate
Tributyl phosphate
Catalog # EHERC17668000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCCOP(=O)(OCCCC)OCCCC
- Inchi: InChI=1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3
- IUPAC: tributyl phosphate
Specifications
Specifications
- Description:Tributyl phosphate
- Cat. No.:EHERC17668000
- Formula:C₁₂H₂₇O₄P
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08
- GHS signal:Danger
- Synonym(s):Tri-n-butyl phosphate;Phosphoric acid tributyl ester
- Molecular weight:266,31
- CAS No.:126-73-8
- Shipping conditions:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



