Order Entry
Italy
Orders LinkContactUsLinkComponent
Mesosulfuron-methyl

Mesosulfuron-methyl

Mesosulfuron-methyl
Catalog # EHERC14913500
Supplier:   EHRENSTORFER
Mesosulfuron-methyl
Catalog # EHERC14913500
Supplier:  EHRENSTORFER
CAS Number:  
Mesosulfuron-methyl
Mesosulfuron-methyl

Mesosulfuron-methyl

CAS Number:  
Price
157,00 €
Log in for availability
1 * 0,1 g
Product Details & Documents
  • SMILES: COC(=O)c1ccc(CNS(=O)(=O)C)cc1S(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2
  • Inchi: InChI=1S/C17H21N5O9S2/c1-29-13-8-14(30-2)20-16(19-13)21-17(24)22-33(27,28)12-7-10(9-18-32(4,25)26)5-6-11(12)15(23)31-3/h5-8,18H,9H2,1-4H3,(H2,19,20,21,22,24)
  • IUPAC: methyl 2-[(4,6-dimethoxypyrimidin-2-yl)carbamoylsulfamoyl]-4-(methanesulfonamidomethyl)benzoate
  • Packing group: III
Specifications

Specifications

  • Description:
    Mesosulfuron-methyl
  • UN No.:
    3077
  • Cat. No.:
    EHERC14913500
  • Formula:
    C₁₇H₂₁N₅O₉S₂
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS09
  • GHS signal:
    Warning
  • Synonym(s):
    AE-F 130060;AE-F 130060-00;Mesomaxx;Mesosulfuron-methyl;Shima
  • Molecular weight:
    503,51
  • CAS No.:
    208465-21-8
  • Shipping conditions:
    RT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.