Hydrocortisone
Hydrocortisone
EHERC14224000
- SMILES: C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
- Inchi: InChI=1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1
- IUPAC: (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
- Description:Hydrocortisone
- Cat. No.:EHERC14224000
- Formula:C₂₁H₃₀O₅
- Storage conditions:20 °C
- Hazard class description:GHS08
- GHS signal:Danger
- Synonym(s):Hydrocortisone;Cortisol;11beta,17,21-Trihydroxy-pregn-4-ene-3,20-dione;Hydrocortisone Hydrogen Succinate Imp. A (EP);Prednisolone Imp. A (EP);Hydrocortisone Acetate Imp. A (Pharmeuropa);17-Hydroxycorticosterone;Acticort;Aeroseb HC;Ala-Cort;Anflam;Anti-inflammatory hormone;CCN 90306A;CaldeCort Spray;Cetacort;Cobadex;Cort-Dome;Cortanal;Cortef;Cortenema;Corticreme;Cortifan;Cortiment;Cortispray;Cortonema;Cortril;Dermacort;Dermocortal;Dermolate;Dihydrocostisone;Dioderm;Domolene-HC;Efcorbin;Efcortelan;Eldecort;Epiderm H;Esiderm H;Evacort;Ficortril;Genacort;HC;Heb-Cort;Hidro-Colisona;Hycort;Hycortol;Hycortole;Hydracort;Hydrasson;Hydro-Adreson;Hydrocort;Hydrocortal;Hydrocorticosterone;Hydrocortisyl;Hydrocortone;Hydroskin;Hytone;Hytone lotion;Incortin-H;Kendall's compound F;Lacticare HC;Medicort;Mildison;NSC 10483;Nutracort;Optef;Otosone-F;Penecort;Permicort;Polcort H;Reichstein's substance M;Scheroson F;Signef;Synacort;Tarcortin;Texacort;Texacort lotion 25;Timocort;Topicort;Traumaide;U 1851;Zenoxone;11beta,17,21-Trihydroxyprogesterone;11beta,17alpha,21-Trihydroxypregn-4-ene-3,20-dione;11beta-Hydroxycortisone;17alpha-Hydroxycorticosterone;4-Pregnene-11beta,17alpha,21-triol-3,20-dione;Hydrocortisone;Hydrocortisone Acetate Impurity A;Hydrocortisone Hydrogen Succinate Impurity A;Prednisolone Impurity A
- Molecular weight:362,46
- CAS No.:50-23-7
- Shipping conditions:RT



