Atrazine-2-hydroxy
Atrazine-2-hydroxy
Catalog # EHERC10333000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCNc1nc(O)nc(NC(C)C)n1
- Inchi: InChI=1S/C8H15N5O/c1-4-9-6-11-7(10-5(2)3)13-8(14)12-6/h5H,4H2,1-3H3,(H3,9,10,11,12,13,14)
- IUPAC: 4-(ethylamino)-6-(propan-2-ylamino)-1,3,5-triazin-2-ol
- Packing group: III
Specifications
Specifications
- Description:Atrazine-2-hydroxy
- UN No.:3077
- Cat. No.:EHERC10333000
- Formula:C₈H₁₅N₅O
- Storage conditions:20 °C
- Hazard class description:GHS08, GHS09
- GHS signal:Warning
- Synonym(s):Atrazine-2-hydroxy
- Molecular weight:197,24
- CAS No.:2163-68-0
- Shipping conditions:RT



