2-Methylcyclohexanone
2-Methylcyclohexanone
Codice catalogo EHERC15085010
Nome del fornitore: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC1CCCCC1=O
- Inchi: InChI=1S/C7H12O/c1-6-4-2-3-5-7(6)8/h6H,2-5H2,1H3
- IUPAC: 2-methylcyclohexan-1-one
- Packing group: III
Specifications
Specifications
- Description:2-Methylcyclohexanone
- UN No.:2297
- Cat. No.:EHERC15085010
- Formula:C₇H₁₂O
- Storage temperature:20 °C
- Hazard class description:GHS02, GHS07
- GHS signal:Warning
- Synonym(s):Cyclohexanone;2-methyl-;2-Methylcyclohexanone;(±)-2-Methylcyclohexanone;2-Methyl-1-cyclohexanone;2-Methylcyclohexane-1-one;NSC 524;α-Methylcyclohexanone
- Molecular weight:112,17
- CAS No.:583-60-8
- Shipping conditions:RT



