Order Entry
Italy
Orders LinkContactUsLinkComponent
Choline ≥98%

Choline ≥98%

Bottle
Catalog # APOSBUP19620-1G
New Product
Supplier:   Apollo Scientific
Choline ≥98%
Catalog # APOSBUP19620-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  123-41-1
Choline ≥98%

Bottle

CAS Number:  123-41-1
Price
37,70 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  2-Hydroxyethyltrimethylammonium Hydroxide, Choline
  • CAS Number:123-41-1
  • Formula:C₅H₁₅NO₂
  • Molecular Weight:121,18 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications