Calcein ≥98%
Bottle
Catalog # APOSBUP17456-100MG
New Product
Supplier: Apollo Scientific
Calcein ≥98%
Catalog # APOSBUP17456-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 1461-15-0
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: Oftasceina, Fluorescein Complexon, Oftasceinum, 2,7-Bis[bis(carboxymethyl)aminomethyl]fluorescein, Oftasceine, 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorexon, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein, NSC 298193
- CAS Number:1461-15-0
- Formula:C₃₀H₂₆N₂O₁₃
- Molecular Weight:622,53 g/mol
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



