Order Entry
Italy
Orders LinkContactUsLinkComponent
 

2-Hydroxy-3-methyl-5-nitropyridine 98%

Supplier:   Apollo Scientific
2-Hydroxy-3-methyl-5-nitropyridine 98%
Supplier:  Apollo Scientific
Certificates
2-Hydroxy-3-methyl-5-nitropyridine 98%

6 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  3-Methyl-5-nitropyridin-2-ol, 3-Methyl-5-nitropyridin-2(1H)-one, 3-Methyl-5-nitro-2-pyridone, 2-Hydroxy-3-methyl-5-nitropyridine
  • CAS Number:21901-34-8
  • Formula:C₆H₆N₂O₃
  • MDL Number:MFCD03095073
  • Molecular Weight:154,12344 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 30,40 €
    30,40 €
    1 * 25 g
       30,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 62,30 €
    62,30 €
    1 * 100 g
       62,30 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 301,00 €
    301,00 €
    1 * 500 g
       301,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 460,00 €
    1 460,00 €
    1 * 2,5 kg
       1 460,00 €

    Specifications:
    New Product
    New Product