Order Entry
Italy
Orders LinkContactUsLinkComponent
Maltose monohydrate ≥98%

Maltose monohydrate ≥98%

Bottle
Catalog # APOSBUP19749-5G
New Product
Supplier:   Apollo Scientific
Maltose monohydrate ≥98%
Catalog # APOSBUP19749-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  6363-53-7
Maltose monohydrate ≥98%

Bottle

CAS Number:  6363-53-7
Price
37,70 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
  • CAS Number:6363-53-7
  • Formula:C₁₂H₂₂₀₁₁·H₂O
  • Molecular Weight:360,31 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.