Dioctyl adipate 98%
Bottle
Codice catalogo APOSOR1000040-5G
New Product
Nome del fornitore: Apollo Scientific
Dioctyl adipate 98%
Codice catalogo APOSOR1000040-5G
Nome del fornitore: Apollo Scientific
New Product
Numero CAS: 123-79-5
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
- CAS Number:123-79-5
- Formula:C₂₂H₄₂O₄
- MDL Number:MFCD00072258
- Molecular Weight:370,57 g/mol
- EINECS:204-652-9
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSOR1000040-5G
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



