Order Entry
Italy
Orders LinkContactUsLinkComponent
 

α-Vitamin E ≥98%

New Product
Supplier:   Apollo Scientific
α-Vitamin E ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
α-Vitamin E ≥98%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  (+)-α-Tocopherol, Vitamin E Alcohol, (R)-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, α-Vitamin E, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E, D(+)-α-Tocopherol
  • CAS Number:59-02-9
  • Formula:C₂₉H₅₀O₂
  • Molecular Weight:430,71 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 49,20 €
    49,20 €
    1 * 50 mg
       49,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 734,00 €
    734,00 €
    1 * 10 g
       734,00 €

    Specifications:
    New Product
    New Product