Order Entry
Italy
Orders LinkContactUsLinkComponent
Trans-Anethole ≥98%

Trans-Anethole ≥98%

Bottle
Catalog # APOSBUP19185-1ML
New Product
Supplier:   Apollo Scientific
Trans-Anethole ≥98%
Catalog # APOSBUP19185-1ML
Supplier:  Apollo Scientific
New Product
CAS Number:  4180-23-8
Trans-Anethole ≥98%

Bottle

CAS Number:  4180-23-8
Price
37,70 €
Log in for availability
1 * 1 mL
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  (E)-1-Methoxy-4-(prop-1-en-1-yl)benzene
  • CAS Number:4180-23-8
  • Formula:C₁₀H₁₂O
  • Molecular Weight:148,2 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.