Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Lithium oxide 99% (by titrimetric analysis)

New Product
Supplier:   Apollo Scientific
Lithium oxide 99% (by titrimetric analysis)
Supplier:  Apollo Scientific
New Product
Certificates
Lithium oxide 99% (by titrimetric analysis)

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Lithium oxide
  • CAS Number:12057-24-8
  • Formula:Li₂O
  • Molecular Weight:29,88 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 23,20 €
    23,20 €
    1 * 5 g
       23,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 60,80 €
    60,80 €
    1 * 25 g
       60,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 205,00 €
    205,00 €
    1 * 100 g
       205,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 724,00 €
    724,00 €
    1 * 500 g
       724,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 3 470,00 €
    3 470,00 €
    1 * 2,5 kg
       3 470,00 €

    Specifications:
    New Product
    New Product