Order Entry
Italy
Orders LinkContactUsLinkComponent
DIVTECH Fluogreen Peptide Delivery Nanoparticles
DIVTECH Fluogreen Peptide Delivery Nanoparticles
Fluogreen peptide delivery nanoparticles (Net-negatively charged peptides; Adherent and suspension cells, organoids, 3D cultures, bacteria, rodents and zebrafish), 1 ml
Catalog # DIVSDIV042F1
New Product
DIVTECH Fluogreen Peptide Delivery Nanoparticles
Catalog # DIVSDIV042F1
New Product
CAS Number:  
DIVTECH Fluogreen Peptide Delivery Nanoparticles
DIVTECH Fluogreen Peptide Delivery Nanoparticles
Fluogreen peptide delivery nanoparticles (Net-negatively charged peptides; Adherent and suspension cells, organoids, 3D cultures, bacteria, rodents and zebrafish), 1 ml
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Peptide-based research demands delivery systems that are both efficient and gentle. DIVERSA’s Peptide delivery nanoparticles are optimised for negatively charged peptides, ensuring stability in biological media and maintaining activity during experiments. Suitable for 2D, 3D, and in vivo studies, they offer a straightforward way to evaluate therapeutic potential. With the fluorescence labeling, researchers can easily monitor uptake and distribution, turning complex delivery studies into streamlined workflows.
  • Easy-to-use formulation
    Non-toxic and biodegradable components
    Optimized for anionic peptides at physiological pH
    Stable in biological media
    Suitable for in vitro and in vivo applications
    Effective in 2D and 3D cell cultures, including immortalise cells, primary cells and organoids
    No specialised equipment required for preparation
    Ideal for internalisation and biodistribution studies
    Trackable by standard fluorescence detection platforms
Peptides are increasingly important as therapeutic candidates but face significant hurdles in delivery due to degradation and poor stability in biological media. DIVERSA’s Peptide delivery nanoparticles are specifically optimised for anionic peptides, forming stable, biocompatible complexes that maintain functionality under physiological conditions.
The kit is suited for both in vitro experiments and in vivo validation, providing researchers with a reproducible and scalable delivery solution. Its compatibility with 2D monolayers, 3D cultures, and organoid systems allows assessment of peptide activity across biologically relevant models.
Fluorescent versions of the kit enable straightforward monitoring of uptake and biodistribution, supporting studies with microscopy, flow cytometry, or plate readers. By offering a user-friendly format and reliable performance, DIVERSA’s Peptide delivery nanoparticles simplify peptide formulation and accelerate the translation of peptide-based therapies from the lab to preclinical testing.
Delivery information: The kit contains one vial with the lipid mix for formulation. Each kit is provided with 1X DIVTECH empty vial for formulation preparation. Includes two pipette tips for 1 ml pipette to support accurate handling. Lot number and storage conditions indicated on every vial and box.
Caution: Products are intended for research use only (RUO).
Specifications
Specifications
  • Description:
    Fluogreen peptide delivery nanoparticles (Net-negatively charged peptides; Adherent and suspension cells, organoids, 3D cultures, bacteria, rodents and zebrafish)
  • Volume:
    1 ml