Order Entry
Italy
Orders LinkContactUsLinkComponent
Tryptone

Tryptone

Tryptone, powder format
Catalog # ICNA091010817
Supplier:   MP Biomedicals
Tryptone
Catalog # ICNA091010817
Supplier:  MP Biomedicals
CAS Number:  
Tryptone
Tryptone

Tryptone, powder format

CAS Number:  
Price
181,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Tryptone is a pancreatic digest of casein, utillised as a source of nitrogen in many culture media for growing bacteria as well as fungi.

  • Lack of detectable carbohydrates

  • An excellent choice for bacterial studies based on fermentation reactions
  • High level of tryptophan makes it useful in the production of indol
  • Used to supplement media in the cultivation of anaerobes (Clostridia and other fermenting organisms, e.g., Lactobacilli)

This preparation supplements media for the cultivation and maintenance of fastidious aerobic and facultative microorganisms such as E. coli and Pseudomonas sp.

Documents

Specifications

Specifications

  • Description:
    Tryptone, powder format
  • Cat. No.:
    ICNA091010817
  • Storage temperature:
    Room temperature (15...30 °C)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.