Proteometer-CV mobile phase A salts
Catalog # NOVIBUF-PCV-001
Supplier: NOVILYTIC
New Product
CAS Number:
Jump to:
Product Details & Documents
5 sealed salt packets specifically formulated for use with Proteometer-CV kit.
Mobile Phase salt packets available for reorder when the Proteometer-CV Reactor, MD Unit, and/or Proteometer-CV reagent are not required
Intended for use with the Proteometer-CV kit.
Ordering information: Reorder in sets of 5 as needed.
Packaging: Supplied in a custom box with instructions for use. Store at room temperature.
Documents
Specifications
Specifications
- Description:Proteometer-CV Mobile Phase A Salt Packets (5X)
Specifications
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..