DAF-FM ≥98% fluorescent probe, for nitric oxide detection
About this item
Highly sensitive, photo-stable fluorescent probe for quantification of low concentrations of nitric oxide (NO). More sensitive to nitric oxide (NO) than DAF-2, More photo-stable and less pH sensitive than DAF-2, Not as cell-permeable as the diacetate derivative, DAF-FM DA , but it does not need to be activated by cytosolic enzymes. Essentially nonfluorescent until it reacts with NO to form a fluorescent benzotriazole.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:254109-20-1
- Molecular Weight:412,34 g/mol
2 Options Available Below
Product Details & Documents
Highly sensitive, photo-stable fluorescent probe for quantification of low concentrations of nitric oxide (NO). More sensitive to nitric oxide (NO) than DAF-2, More photo-stable and less pH sensitive than DAF-2, Not as cell-permeable as the diacetate derivative, DAF-FM DA , but it does not need to be activated by cytosolic enzymes. Essentially nonfluorescent until it reacts with NO to form a fluorescent benzotriazole.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:254109-20-1
- Molecular Weight:412,34 g/mol
Specifications for Product Family
Specification Test Results
- Constituents::100% 7-amino-2',7'-difluoro-3',6'-dihydroxy-6-(methylamino)spiro[2-benzofuran-3,9'-xanthene]-1-one
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



