Order Entry
Italy
Orders LinkContactUsLinkComponent
Alcian blue <gt/>50% Cationic dye

Alcian blue 50% Cationic dye

Codice catalogo ABCAAB145250-5
Marketsource
Nome del fornitore:   Abcam
Alcian blue <gt/>50% Cationic dye
Codice catalogo ABCAAB145250-5
Nome del fornitore:  Abcam
Marketsource
Numero CAS:  33864-99-2
Alcian blue 50% Cationic dye
Numero CAS:  33864-99-2
Price
205,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Cationic dye. Forms insoluble complexes with glycosaminoglycans in solution. Stains interphase nuclei, especially mitotic figures.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:33864-99-2
  • Molecular Weight:1298,9 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Alcian Blue
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.