Daidzein ≥99% Phytoestrogen
Product Details & Documents
Phytoestrogen, and soy isoflavone. Analog of genistein. Various biological actions, including a weak estrogenic / antiestrogenic effect by binding to the nuclear oestrogen receptor.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:486-66-8
- Molecular Weight:254,24 g/mol
Specifications
Specification Test Results
- Constituents::100% Daidzein
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



