Order Entry
Italy
ContactUsLinkComponent
NANOCOLOR® Photometric Tube Test Kit for Bitter Units
NANOCOLOR® Photometric Tube Test Kit for Bitter Units
Catalog # MANA918172
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Test Kit for Bitter Units
Catalog # MANA918172
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Isooctane extraction
  • Pk:
    1 SET
  • Description:
    NANOCOLOR® Bitter units, test no. 1-72
  • Dimensions:
    160×110×119 mm (including packaging)
  • Model:
    MANA918172
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    15…25 °C
  • Shelf life (months):
    12
  • Weight:
    367.8 g (including packaging)
  • For unit:
    UV/VIS II; VIS II; 500 D; PF-12Plus; Advance
  • Tests per kit:
    20
  • Range:
    2,0 - 80,0 (BU)

Specifications

About this item

Photometric test kit for the determination of bitterness units in beer and wort samples.

  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes 16 mm (OD) tubes for easy pipetting

The test kit is evaluated in 10 mm quartz glass cells. It is preprogrammend on the NANOCOLOR® UV/VIS II photometer and also compatible with METTLER TOLEDO Easy UV. Programming data for other UV spectrophotometers are available.

Sea water analysis: No
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR UV/VIS II

Delivery: Rugged box with 20 test tubes.