Order Entry
Italy
ContactUsLinkComponent
NANOCOLOR® Photometric Tube Test Kit for Potassium
NANOCOLOR® Photometric Tube Test Kit for Potassium
Catalog # MANA985045
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Test Kit for Potassium
Catalog # MANA985045
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Potassium tetraphenylborate (turbidity)
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Potassium 50, test no. 0-45
  • Dimensions:
    160×110×119 mm (including packaging)
  • Model:
    MANA985045
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    15…25 °C
  • Shelf life (months):
    24
  • Weight:
    386.4 g (including packaging)
  • For unit:
    UV/VIS II; VIS II; 500 D; PF-12Plus; Advance
  • Tests per kit:
    20
  • Range:
    2 - 50 mg/L (K⁺)

Specifications

About this item

Photometric test kit for the determination of potassium in water samples.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting

The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.

Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II

Delivery: Rugged box with 20 test tubes.