Order Entry
Italy
ContactUsLinkComponent
NANOCOLOR® Photometric Tube Test Kit for Carbonate Hardness
NANOCOLOR® Photometric Tube Test Kit for Carbonate Hardness
Catalog # MANA985015
Nome del fornitore:  MACHEREY-NAGEL
Numero CAS:  
undefined
NANOCOLOR® Photometric Tube Test Kit for Carbonate Hardness
Catalog # MANA985015
Nome del fornitore:  MACHEREY-NAGEL
Numero CAS:  

Specifiche

  • Chemical method:
    Bromophenol blue
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Carbonate hardness 15, test no. 0-15
  • Dimensions:
    160×110×119 mm (Including packaging)
  • Model:
    MANA985015
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    12
  • Weight:
    384 g (Including packaging)
  • For unit:
    UV/VIS II;VIS II;500 D;PF-12Plus;Advance;MT Easy VIS;MT Easy UV
  • Tests per kit:
    20
  • Range:
    1,0 - 18,0 - °e; 0,4 - 5,4 mmol/L (H⁺); 20 - 270 mg/L (CaCO₃)

Specifiche

Informazioni su questo articolo

Photometric test kit for the determination of carbonate hardness in water samples.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents

  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (ext.Ø) tubes for easy pipetting

The test kit is evaluated in 16 mm (ext.Ø) tubes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.

Sea water analysis: Yes

Hazardous material: No

Remark: Measuring range on NANOCOLOR VIS II

Sludge reagent set: No

Digestion set: No

NanOx Metal: No

NanOx N: No

Delivery: Rugged box with 20 test tubes. Sufficient for 20 tests.