Order Entry
Italy
ContactUsLinkComponent
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
New Product
Certificates
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
New Product
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99.8 %
  • Acetone and Aldehyde (as(CH3)2CO):
    max. 0.005 %
  • Acid and Chloride:
    passes test
  • Color (APHA):
    ≤ 10
  • Free Chlorine:
    passes test
  • Preservative (C2H5OH):
    about 0.75%
  • Residue after Evaporation:
    max. 0.001 %
  • Substances Darkened by H2SO4:
    passes test
  • Suitability for Use with Dithizone Test:
    passes test
  • Aluminium (Al):
    max. 0.5
  • Barium (Ba):
    max. 0.1
  • Boron (B):
    max. 0.02
  • Cadmium (Cd):
    max. 0.05
  • Calcium (Ca):
    max. 0.5
  • Chromium (Cr):
    max. 0.02
  • Cobalt (Co):
    max. 0.02
  • Copper (Cu):
    max. 0.02
  • Iron (Fe):
    max. 0.1
  • Lead (Pb):
    max. 0.05
  • Magnesium (Mg):
    max. 0.1
  • Manganese (Mn):
    max. 0.02
  • Nickel (Ni):
    max. 0.02
  • Tin (Sn):
    max. 0.1
  • Zinc (Zn):
    max. 0.1

Specification Test Results

Related Information

Frequently Bought Together