Liquid Ends for Liquid Handling Station
Supplier: Brand
Certificates
About this item
The basis for the pipette modules are components from the familiar Transferpette® S.
Autoclavable at 121 °C for 20 minutes
Three single-channel Liquid Ends (SC) and three 8-channel Liquid Ends (MC) are available for contact-free liquid delivery. The volume testing of the pipette modules is carried out according to ISO 8655 part 6.
Related Information
Liquid Handling Station.pdf
Application Notes
TDS Liquid Handling Station.pdf
Datasheet
Frequently Bought Together