Order Entry
Italy
ContactUsLinkComponent
Endeavor™ Orbital Shakers
Endeavor™ Orbital Shakers
Supplier:  OHAUS
New Product
Certificates
undefined
Endeavor™ Orbital Shakers
Supplier:  OHAUS
New Product
Specifications
  • Catalog No:
  • OHAU30770916
  • OHAU30770915
  • Model:
  • e-E51LD0420
  • e-E51LD0403
  • Speed:
  • 50 - 300 min⁻¹
  • 100 - 800 min⁻¹
  • Max. capacity:
  • 4 kg (up to 3×1 L flasks)
  • 4 kg (up to 3×1 L flasks)
  • Orbit:
  • 20 mm
  • 3 mm