Mouse ALT ELISA Kit
Supplier: Abcam
New Product
Certificates
About this item
Mouse ALT ELISA Kit is a single-wash, 90-minute SimpleStep ELISA® used to quantify mouse alanine aminotransferase (ALT) with a sensitivity of 4 pg/ml. The assay follows a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric Sandwich ELISA – 450 nm readout, compatible with any standard plate reader - Different formats for different needs – 384-well format available for high-throughput applications - Broad sample compatibility – works with serum, plasma - Cited in over 15 publications.
Frequently Bought Together