Order Entry
Italy
ContactUsLinkComponent
17 OH Progesterone ELISA Kit
17 OH Progesterone ELISA Kit
Catalog # ABCAAB108668-96
Supplier:  Abcam
CAS Number:  
undefined
17 OH Progesterone ELISA Kit
Catalog # ABCAAB108668-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Target protein:
    Progesterone 17 Alpha OH
  • Description:
    17 OH Progesterone ELISA kit
  • Sample type:
    Citrate plasma, heparin plasma, serum
  • Detection method:
    Colorimetric
  • Storage temperature:
    4 °C
  • Sensitivity:
    0,05 ng/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

17 OH Progesterone ELISA kit is a Competitive ELISA kit for the measurement of 17 OH Progesterone in Citrate plasma, heparin plasma, serum samples.