Melatonin ELISA kit (salivary)
Catalog # ABCAAB283258-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Melatonin ELISA kit (salivary)
- Tests per kit:96 tests
Specifications
About this item
Melatonin ELISA kit (salivary) is a Competitive ELISA kit for the measurement of Melatonin (salivary) in Human in Saliva samples.