Canine HGF ELISA Kit
Catalog # ABCAAB273174-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Canine HGF ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Canine HGF ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Canine HGF in Dog in Plasma, cell culture supernatant, serum samples.