Order Entry
Italy
Orders LinkContactUsLinkComponent
Mouse PD-1H ELISA Kit

Mouse PD-1H ELISA Kit

Mouse PD-1H ELISA kit, 96 tests
Catalog # ABCAAB231933-96
Supplier:   Abcam
Mouse PD-1H ELISA Kit
Catalog # ABCAAB231933-96
Supplier:  Abcam
CAS Number:  
Mouse PD-1H ELISA Kit
Mouse PD-1H ELISA Kit

Mouse PD-1H ELISA kit, 96 tests

CAS Number:  
Price
800,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse PD-1H ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse PD-1H with a sensitivity of 44,3 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.

Specifications

Specifications

  • Description:
    Mouse PD-1H ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Tissue Extracts, Heparin Plasma, Citrate plasma, Serum, Cell Lysate, EDTA Plasma
  • Detection range:
    390,63 - 25000 pg/ml
  • Storage temperature:
    +4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB231933-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    44,3 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    VISTA
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.