Order Entry
Italy
Orders LinkContactUsLinkComponent
Human Cripto ELISA Kit

Human Cripto ELISA Kit

Human Cripto ELISA kit, 96 tests
Catalog # ABCAAB245719-1X96
Supplier:   Abcam
Human Cripto ELISA Kit
Catalog # ABCAAB245719-1X96
Supplier:  Abcam
CAS Number:  
Human Cripto ELISA Kit
Human Cripto ELISA Kit

Human Cripto ELISA kit, 96 tests

CAS Number:  
Price
744,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Human Cripto ELISA kit is a single-wash 90-min Simplestep used to quantify Human Cripto with a sensitivity of 9,6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Specifications

Specifications

  • Description:
    Human Cripto ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
  • Detection range:
    26,56 - 1700 pg/ml
  • Storage temperature:
    4 ℃
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB245719-1X96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    9,6 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Cripto1/CRIPTO
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.