Order Entry
Italy
ContactUsLinkComponent
Human/Monkey MCP4 Antibody Pair - BSA and Azide Free
Human/Monkey MCP4 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB244169-10X96
Supplier:  Abcam
CAS Number:  
undefined
Human/Monkey MCP4 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB244169-10X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Target protein:
    MCP4
  • Description:
    Human/Monkey MCP4 antibody pair - BSA and azide free
  • Detection range:
    3,9 - 250 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    10×96 tests

Specifications

About this item

Human/Monkey MCP4 antibody pair - BSA and Azide free.