Mouse Fibrinogen ELISA Kit
Catalog # ABCAAB108843-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Fibrinogen ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Fibrinogen ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Fibrinogen in Mouse in Urine, Plasma samples.