Mouse Vitronectin ELISA Kit
Catalog # ABCAAB272778-96TST
Nome del fornitore: Abcam
New Product
Numero CAS:
Specifiche
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:Vitronectin/S-Protein
- Description:Mouse Vitronectin ELISA kit
- Detection range:0,1 - 100 ng/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:96 tests
Specifiche
Informazioni su questo articolo
Mouse Vitronectin ELISA kit is a Quantitative ELISA kit for the measurement of Mouse Vitronectin in Mouse in Citrate plasma, EDTA plasma, Heparin plasma, Other biological fluids samples.