Order Entry
Italy
Orders LinkContactUsLinkComponent
Human Complement C6 ELISA Kit

Human Complement C6 ELISA Kit

Human Complement C6 ELISA kit, 96 tests
Catalog # ABCAAB230934-96
Supplier:   Abcam
Human Complement C6 ELISA Kit
Catalog # ABCAAB230934-96
Supplier:  Abcam
CAS Number:  
Human Complement C6 ELISA Kit
Human Complement C6 ELISA Kit

Human Complement C6 ELISA kit, 96 tests

CAS Number:  
Price
795,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human Complement C6 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Complement C6 with a sensitivity of 71 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.

Specifications

Specifications

  • Description:
    Human Complement C6 ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Saliva, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
  • Detection range:
    0,2 - 15 ng/ml
  • Storage temperature:
    +4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB230934-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    71 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    C6
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.