Phospho-ATF2 (T69/71) ELISA Kit
Catalog # ABCAAB279737-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Phospho-ATF2 (T69/71) ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Phospho-ATF2 (T69/71) ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phospho-ATF2 (T69/71) in Human in Cell Lysate samples.